MilliporeSigma
  • Inhibition of monoamine oxidase by C5-substituted phthalimide analogues.
Inhibition of monoamine oxidase by C5-substituted phthalimide analogues.
Bioorganic & medicinal chemistry (2011-07-23)
Clarina I Manley-King, Jacobus J Bergh, Jacobus P Petzer
ABSTRACT
Literature reports that isatin as well as C5- and C6-substituted isatin analogues are reversible inhibitors of human monoamine oxidase (MAO) A and B. In general, C5- and C6-substitution of isatin leads to enhanced binding affinity to both MAO isozymes compared to isatin and in most instances result in selective binding to the MAO-B isoform. Crystallographic and modeling studies suggest that the isatin ring binds to the substrate cavities of MAO-A and -B and is stabilized by hydrogen bond interactions between the NH and the C2 carbonyl oxygen of the dioxoindolyl moiety and water molecules present in the substrate cavities of MAO-A and -B. Based on these observations and the close structural resemblances between isatin and its phthalimide isomer, a series of phthalimide analogues were synthesized and evaluated as MAO inhibitors. While phthalimide and N-aryl-substituted phthalimides were found to be weak MAO inhibitors, phthalimide homologues containing C5 substituents were potent reversible inhibitors of recombinant human MAO-B with IC(50) values ranging from 0.007 to 2.5 μM and moderately potent reversible inhibitors of recombinant human MAO-A with IC(50) values ranging from 0.22 to 9.0 μM. By employing molecular docking the importance of hydrogen bonding between the active sites of MAO-A and -B and the phthalimide inhibitors are highlighted.
Sigma-Aldrich
Phthalimide, ≥99%
Sigma-Aldrich
Isatin, 97%