MilliporeSigma
  • Intellectual capital-based performance improvement: a study in healthcare sector.
Intellectual capital-based performance improvement: a study in healthcare sector.
BMC health services research (2021-01-21)
Simona Alfiero, Valerio Brescia, Fabrizio Bert
ABSTRACT
Knowledge resources are in most productive sectors distinctive in terms of competitiveness. Still, in the health sector, they can have an impact on the health of the population, help make the organisations more efficient and can help improve decision-making processes. The purpose of this paper is to investigate the Intellectual Capital impact on healthcare organization' performance in the Italian healthcare system. The theoretical framework linked to intellectual Capital in the health sector and the performance evaluation related to efficiency supports the analysis carried out in two stages to determine the right placement of resources and the exogenous variables that influence performance level. The evaluation of the impact of the ICs on performance is determined through the Data envelopment analysis. The incidence of the exogenous variables has been established through linear regression. Empirical results in Italy show some IC components influence organization 'performance (Essential Levels of Assistance) and could be used for defining the policy of allocation of resources in healthcare sector. The efficiency of 16 regions considered in 2016 based on Slack-Based-Model constant returns-to-scale (SBM-CRS) and Slack-Based-Model variable returns-to-scale (SBM-VRS) identifies a different ability to balance IC and performance. Current healthcare expenditure and the number of residents is correlated with the identified efficiency and performance levels. This paper embeds an innovative link between healthcare performance, in term of efficiency and IC which aligns resource management with future strategy. The study provides a new decision-making approach.
Sigma-Aldrich
N,N′-Bis(hydroxymethyl)urea, ≥95%