MilliporeSigma
  • Rab22a-NeoF1 fusion protein promotes osteosarcoma lung metastasis through its secretion into exosomes.
Rab22a-NeoF1 fusion protein promotes osteosarcoma lung metastasis through its secretion into exosomes.
Signal transduction and targeted therapy (2021-02-12)
Li Zhong, Dan Liao, Jingjing Li, Wenqiang Liu, Jingxuan Wang, Cuiling Zeng, Xin Wang, Zhiliang Cao, Ruhua Zhang, Miao Li, Kuntai Jiang, Yi-Xin Zeng, Jianhua Sui, Tiebang Kang
ABSTRACT
It remains unknown for decades how some of the therapeutic fusion proteins positive in a small percentage of cancer cells account for patient outcome. Here, we report that osteosarcoma Rab22a-NeoF1 fusion protein, together with its binding partner PYK2, is sorted into exosomes by HSP90 via its KFERQ-like motif (RVLFLN142). The exosomal Rab22a-NeoF1 fusion protein facilitates the pulmonary pre-metastatic niche formation by recruiting bone marrow-derived macrophages. The exosomal PYK2 activates RhoA in its negative recipient osteosarcoma cells and induces signal transducer and activator of transcription 3 activation in its recipient macrophages to increase M2 phenotype. Consequently, lung metastases of its recipient osteosarcoma cells are promoted by this exosomal Rab22a-NeoF1 fusion protein, and this event can be targeted by disrupting its interaction with PYK2 using a designed internalizing RGD peptide.
Sigma-Aldrich
MISSION® esiRNA, targeting human PDCD6IP
Sigma-Aldrich
MISSION® esiRNA, targeting human TSG101
Sigma-Aldrich
MISSION® esiRNA, targeting human PTK2B
Sigma-Aldrich
MISSION® esiRNA, targeting human RAB11A
Sigma-Aldrich
MISSION® esiRNA, targeting human RAB27A