MilliporeSigma
  • Inhibition of plasma kallikrein mitigates experimental hypertension-enhanced cerebral hematoma expansion.
Inhibition of plasma kallikrein mitigates experimental hypertension-enhanced cerebral hematoma expansion.
Brain research bulletin (2021-02-09)
Jie Wang, Min Xia, Xiaoqin Tang, Zhengcai Jia, Chengcheng Li, Mingxi Li, Yi Yin, Chao Guo, Jiantao Shi, Xin Liu, Weixiang Chen, Tunan Chen, Hua Feng
ABSTRACT
Hematoma expansion (HE) aggravates brain injury after intracerebral hemorrhage (ICH) and hypertension is a key contributor to HE. Plasma kallikrein (PK) is involved in hemorrhagic transformation in ischemic stroke mice. This study was conducted to explore the role of PK in HE in hypertensive ICH. Hypertension was achieved by continuous infusion of angiotensin II (Ang II) with an osmotic pump in C57BL/6 mice. ICH was achieved by stereotactic intrastriatal injection of blood. PK-specific antibody and platelet glycoprotein VI (GPVI) agonists were administered to intervene in hematoma expansion. The hematoma volume was indicated by the erythrocyte components hemoglobin and carbonic anhydrase-1 in the ipsilateral brain hemisphere. Ang II-induced hypertensive mice showed enhanced hematoma expansion and worsened neurologic deficits after ICH modeling. Moreover, intrastriatal injection of blood from Ang II-treated mice into normal mice increased the area of secondary hemorrhage more than blood from untreated mice. Mechanistically, elevated PK was found in Ang II-infused mice whereas, inhibition of PK and administration of the GPVI agonist convulxin decreased hematoma expansion and improved neurologic deficits after ICH. These findings suggest that PK inhibition and GPVI agonist treatment might serve as potential methods to intervene in HE after ICH.
Sigma-Aldrich
Human PSA-total ELISA Kit, for serum, plasma, cell culture supernatant and urine