MilliporeSigma
  • Inhibition of TLR2/TLR4 alleviates the Neisseria gonorrhoeae infection damage in human endometrial epithelial cells via Nrf2 and NF-Kβsignaling.
Inhibition of TLR2/TLR4 alleviates the Neisseria gonorrhoeae infection damage in human endometrial epithelial cells via Nrf2 and NF-Kβsignaling.
Journal of reproductive immunology (2020-09-21)
Yun Yang, Shasha Liu, Jixiao Liu, Na Ta
ABSTRACT
Neisseria gonorrhoeae (N.g) is Gram-negative bacteria and can lead to endometritis in female. Toll-like receptors regulate immune response in various diseases. However, the roles of TLR2 and TLR4 in. Neisseria gonorrhoeae-induced infection damage in human endometrial epithelia were investigated. hEECs were infected with N.g (MOI 10 and 100) and cell viability and apoptosis were measured by CCK8 and flow cytometry assays in both infected groups with the uninfected normal hEECs as negative control. TLR2/TLR4 proteins were measured by ELISA method. Pro-inflammatory markers NLRP3, PGES (PGE2) and TNF-α were assessed by RT-qPCR (mRNA expression) and Elisa (protein concentrations). Transfection assays were performed to up- or down- regulate expression of TLR2 and TLR4 so as to study the functions of TLR2/TLR4 in. N.g-infected hEECs, followed by apoptosis and inflammation assessment. Similarly, we explored the interactions between TLR2/TLR4 and Nrf2/NF-κB/p65 by knocking down TLR2/TLR4 to detect the signaling and further regulating the signaling to evaluate TLR2/ TLR4, apoptosis and inflammation in cells. N.g suppressed cell viabilities and induced cell apoptosis and inflammation. TLR2/TLR4 downregulation inhibited the infection damage. Nrf2 was activated while NF-κB/p65 was depleted as TLR2/ TLR4 was knocked down. Activation of Nrf2 and inhibition of NF-κB resulted in decrease of TLR2/TLR4, which could retard apoptosis and inflammation induced by N.g infection. TLR2/TLR4 depletion could alleviate the N.g-infected hEECs via Nrf2/NF-kB signaling, suggesting that TLR2/TLR4 inhibitors might serve as a treatment to reduce N.g infection in human endometrial epithelia.
Sigma-Aldrich
Human TLR2 / Toll-like Receptor 2 ELISA Kit
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder