MilliporeSigma
  • Loading, release profile and accelerated stability assessment of monoterpenes-loaded solid lipid nanoparticles (SLN).
Loading, release profile and accelerated stability assessment of monoterpenes-loaded solid lipid nanoparticles (SLN).
Pharmaceutical development and technology (2020-03-25)
Aleksandra Zielińska, Nuno R Ferreira, Agnieszka Feliczak-Guzik, Izabela Nowak, Eliana B Souto
ABSTRACT
Glycerol monostearate solid lipid nanoparticles (SLN) were produced by hot high-pressure homogenization technique to load alpha-pinene, citral, geraniol or limonene. SLN were composed of 1 wt.% monoterpene, 4 wt.% of Imwitor® 900K as a solid lipid and 2.5 wt.% of Poloxamer188 as a surfactant. Empty SLN consisted of 5 wt.% of Imwitor® 900K and 2.5 wt.% of Poloxamer188. The mean particles size (Z-Ave) and polydispersity index (PDI) of SLN were analyzed by dynamic light scattering (DLS), while the zeta potential (ZP) of each formulation were measured by electrophoretic light scattering. LUMiSizer® was applied to calculate the velocity distribution in the centrifugal field and instability index. Drug release profile from SLN was analyzed using Franz cell diffusion cells assayed by UV-Vis spectrophotometry, whereas the gas chromatography technique was applied to determine the encapsulation parameters of volatile monoterpenes. The matrix state, polymorphism and phase behavior of SLN were studied by X-ray diffraction (XRD, low and wide angles) and differential scanning calorimetry (DSC). Selected monoterpenes were successfully loaded in glycerol monostearate SLN. A burst release profile within the first 15 min was observed for all formulations, being the modified release profile dependent on the type of monoterpene and on the encapsulation efficiency.
Sigma-Aldrich
α-Pinene, 98%
Sigma-Aldrich
Citral, 95%