MilliporeSigma
  • Urethane Dimethacrylate Influences the Cariogenic Properties of Streptococcus Mutans.
Urethane Dimethacrylate Influences the Cariogenic Properties of Streptococcus Mutans.
Materials (Basel, Switzerland) (2021-03-07)
Kyungsun Kim, Jeong Nam Kim, Bum-Soon Lim, Sug-Joon Ahn
ABSTRACT
Concerns regarding unbound monomers in dental composites have increased with the increased usage of these materials. This study assessed the biological effects of urethane dimethacrylate (UDMA), a common monomer component of dental composite resins, on the cariogenic properties of Streptococcus mutans. Changes in the growth rate, biofilm formation, interaction with saliva, surface hydrophobicity, adhesion, glucan synthesis, sugar transport, glycolytic profiles, and oxidative- and acid-stress tolerances of S. mutans were evaluated after growing the cells in the presence and absence of UDMA. The results indicated that UDMA promotes the adhesion of S. mutans to the underlying surfaces and extracellular polysaccharide synthesis, leading to enhanced biofilm formation. Furthermore, UDMA reduced the acid tolerance of S. mutans, but enhanced its tolerance to oxidative stress, thus favoring the early stage of biofilm development. UDMA did not significantly affect the viability or planktonic growth of cells, but diminished the ability of S. mutans to metabolize carbohydrates and thus maintain the level of intracellular polysaccharides, although the tendency for sugar transport increased. Notably, UDMA did not significantly alter the interactions of bacterial cells with saliva. This study suggests that UDMA may potentially contribute to the development of secondary caries around UDMA-containing dental materials by prompting biofilm formation, enhancing oxidative tolerance, and modulating carbon flow.
Sigma-Aldrich
Phenazine methosulfate
Sigma-Aldrich
Diurethane dimethacrylate, mixture of isomers, contains 225 ppm±25 ppm BHT as inhibitor, ≥97%
BRAND® 96-well microplate, U-bottom, round bottom, non-sterile