MilliporeSigma
  • 2-Hydroxy-4-methoxybenzaldehyde from Hemidesmus indicus is antagonistic to Staphylococcus epidermidis biofilm formation.
2-Hydroxy-4-methoxybenzaldehyde from Hemidesmus indicus is antagonistic to Staphylococcus epidermidis biofilm formation.
Biofouling (2020-06-27)
Arunachalam Kannappan, Ravindran Durgadevi, Ramanathan Srinivasan, Ricardo José Lucas Lagoa, Issac Abraham Sybiya Vasantha Packiavathy, Shunmugiah Karutha Pandian, Arumugam Veera Ravi
ABSTRACT
Staphylococcus epidermidis (SE) is an opportunistic nosocomial pathogen that accounts for recalcitrant device-related infections worldwide. Owing to the growing interest in plants and their secondary metabolites targeting bacterial adhesion, this study was intended to uncover the anti-biofilm potential of Hemidesmus indicus and its major constituent 2-hydroxy-4-methoxybenzaldehyde (HMB) against SE. The minimum biofilm inhibitory concentration (MBIC) of H. indicus root extract and HMB were found to be 500 and 250 µg ml-1, respectively. The results of time-dependent biofilm inhibition and mature biofilm disruption assays confirmed that HMB targets initial cell adhesion. Furthermore, interference by HMB in the expression of adhesin genes (icaA, aap and bhp) and biofilm components was associated with an increased susceptibility of SE to oxidative stress and antibiotics. To conclude, this study reports for the first time HMB as a potential drug against SE biofilms.
Sigma-Aldrich
2-Hydroxy-4-methoxybenzaldehyde, 98%