MilliporeSigma
  • Biofilm interfacial acidity evaluation by pH-Responsive luminescent nanoparticle films.
Biofilm interfacial acidity evaluation by pH-Responsive luminescent nanoparticle films.
Biosensors & bioelectronics (2020-10-30)
Padryk Merkl, Marie-Stephanie Aschtgen, Birgitta Henriques-Normark, Georgios A Sotiriou
ABSTRACT
Biofilms are dense bacterial colonies that may adhere to the surfaces of medical devices and are major contributors to infections. These colonies are characterized by a self-produced matrix of extracellular polymeric substances (EPS). Bacterial biofilms are difficult to treat with the commonly used antibiotics partially because of their poor diffusion through the EPS and therefore require new targeted strategies to effectively fight them. Biofilms may produce an acidic microenvironment which can be exploited to design such targeted treatment strategies. However, there is currently a lack of high-throughput ways to determine the acidity of biofilms at their interface with the medical device. Here, a novel all-inorganic pH responsive system is developed from luminescent carbonated hydroxyapatite nanoparticles doped with Eu3+ ions which can determine the biofilm acidity fluorometrically due to carbonate removal in acidic environments that directly affects the nanoparticle luminescence. The pH responsive nanoparticles are in-situ deposited during their production onto substrates on which a variety of clinically-relevant biofilms are grown. The acidity of their interfacial (micro)environment depends on the bacterial species and strain even when differences in biofilm biomass are considered.
Sigma-Aldrich
Tributyl phosphate, ≥99%