MilliporeSigma
  • Risk assessment of the chiral pesticide fenamiphos in a human model: Cytochrome P450 phenotyping and inhibition studies.
Risk assessment of the chiral pesticide fenamiphos in a human model: Cytochrome P450 phenotyping and inhibition studies.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2020-11-01)
Nayara Cristina Perez de Albuquerque, Daniel Blascke Carrão, Maísa Daniela Habenschus, Franciele Saraiva Fonseca, Rodrigo Moreira da Silva, Norberto Peporine Lopes, Bruno Alves Rocha, Fernando Barbosa Júnior, Anderson Rodrigo Moraes de Oliveira
ABSTRACT
Fenamiphos (FS) is a chiral organophosphate pesticide that is used to control nematodes in several crops. Enantioselective differences may be observed in FS activity, bioaccumulation, metabolism, and toxicity. Humans may be exposed to FS through occupational and chronic (food, water, and environmental) exposure. FS may cause undesirable CYP450 pesticide-drug interactions, which may impact human health. Here, the CYP450 isoforms involved in enantioselective FS metabolism were identified, and CYP450 inhibition by rac-FS, (+)-FS, and (-)-FS was evaluated to obtain reliable information on enantioselective FS risk assessment in humans. CYP3A4 and CYP2E1 metabolized FS enantiomers, and CYP2B6 may participate in rac-FS metabolism. In addition, rac-FS, (+)-FS, and (-)-FS were reversible competitive CYP1A2, CYP2C19, and CYP3A4/5 inhibitors. High stereoselective inhibition potential was verified; rac-FS and (-)-FS strongly inhibited and (+)-FS moderately inhibited CYP1A2. Stereoselective differences were also detected for CYP2C19 and CYP3A4/5, which were strongly inhibited by rac-FS, (+)-FS, and (-)-FS. Our results indicated a high potential for CYP450 drug-pesticide interactions, which may affect human health. The lack of stereoselective research on the effect of chiral pesticides on the activity of CYP450 isoforms highlights the importance of assessing the risks of such pesticides in humans.
Product Description
Supelco
Fenamiphos-sulfoxide, PESTANAL®, analytical standard
Sigma-Aldrich
Nifedipine, ≥98% (HPLC), powder
Sigma-Aldrich
Quinidine, crystallized, ≥98.0% (dried material, NT)