MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Synthesis and anticonvulsant activity of new 1-[2-oxo-2-(4-phenylpiperazin-1-yl)ethyl]pyrrolidine-2,5-diones.
Synthesis and anticonvulsant activity of new 1-[2-oxo-2-(4-phenylpiperazin-1-yl)ethyl]pyrrolidine-2,5-diones.
Bioorganic & medicinal chemistry letters (2011-08-31)
Krzysztof Kamiński, Sabina Rzepka, Jolanta Obniska
ABSTRACT
Twenty-two new 1-[2-oxo-2-(4-phenylpiperazin-1-yl)ethyl]pyrrolidine-2,5-diones were synthesized and tested for anticonvulsant activity. Initial anticonvulsant screening was performed using standard maximal electroshock (MES) and subcutaneous pentylenetetrazole (scPTZ) screens in mice. Several compounds were tested additionally in the 6-Hz psychomotor seizure model. The neurotoxicity was determined applying the rotarod test. Excluding one compound, all other molecules were found to be effective in at least one seizure model. The most active were 1-(2-oxo-2-{4-[3-(trifluoromethyl)phenyl]piperazin-1-yl}ethyl)pyrrolidine-2,5-dione (14), 1-{2-[4-(4-chlorophenyl)piperazin-1-yl]-2-oxoethyl}-3-methylpyrrolidine-2,5-dione (17), 1-{2-[4-(4-chlorophenyl)piperazin-1-yl]-2-oxoethyl}-3,3-dimethylpyrrolidine-2,5-dione (23) and 3,3-dimethyl-1-(2-oxo-2-{4-[3-(trifluoromethyl)phenyl]piperazin-1-yl}ethyl)pyrrolidine-2,5-dione (26). These compounds showed high activity in the 6-Hz psychomotor seizure test as well as were active in the maximal electroshock and subcutaneous pentylenetetrazole (14 and 23) screens. Initial SAR studies for anticonvulsant activity have been discussed.
Sigma-Aldrich
5,5-Diphenylhydantoin, ≥98%
Sigma-Aldrich
1-Phenylpiperazine, ≥97.0%
Sigma-Aldrich
Ethosuximide