MilliporeSigma
  • Electrophoretic Deposition of Aged and Charge Controlled Colloidal Copper Sulfide Nanoparticles.
Electrophoretic Deposition of Aged and Charge Controlled Colloidal Copper Sulfide Nanoparticles.
Nanomaterials (Basel, Switzerland) (2021-01-13)
Yoonsu Park, Hyeri Kang, Wooseok Jeong, Hyungbin Son, Don-Hyung Ha
ABSTRACT
Colloidal nanoparticles (NPs) have been recently spotlighted as building blocks for various nanostructured devices. Their collective properties have been exhibited by arranging them on a substrate to form assembled NPs. In particular, electrophoretic deposition (EPD) is an emerging fabrication method for such nanostructured films. To maximize the benefits of this method, further studies are required to fully elucidate the key parameters that influence the NP deposition. Herein, two key parameters are examined, namely: (i) the aging of colloidal NPs and (ii) the charge formation by surface ligands. The aging of Cu2-xS NPs changes the charge states, thus leading to different NP deposition behaviors. The SEM images of NP films, dynamic light scattering, and zeta potential results demonstrated that the charge control and restoration of interparticle interactions for aged NPs were achieved via simple ligand engineering. The charge control of colloidal NPs was found to be more dominant than the influence of aging, which can alter the surface charges of the NPs. The present results thus reveal that the charge formation on the colloidal NPs, which depends on the surface ligands, is an important controllable parameter in EPD.
Sigma-Aldrich
Di-tert-butyl disulfide, 97%