MilliporeSigma
  • CFL2 is an essential mediator for myogenic differentiation in C2C12 myoblasts.
CFL2 is an essential mediator for myogenic differentiation in C2C12 myoblasts.
Biochemical and biophysical research communications (2020-11-15)
Mai Thi Nguyen, Kyung-Ho Min, Dough Kim, Seung-Yoon Park, Wan Lee
ABSTRACT
CFL2, a skeletal muscle-specific member of the actin depolymerizing factor/cofilin protein family, is known to be involved in the regulation of actin filament dynamics. Although the impact of CFL2 has been studied in human myopathy, its functional contribution to myogenic differentiation, in terms of its effects on cell proliferation, cell cycle, and myogenic factor modulation, remains largely unknown. Here, we report that CFL2 is required for the myogenic differentiation of C2C12 myoblasts by regulating proliferation and myogenic transcription factors expressions. CFL2 expression was induced during myogenic progression, and its knockdown by siRNA in myoblasts enhanced phalloidin staining, indicating increased filamentous actin formation. Interestingly, CFL2 depletion stimulated cell proliferation and induced a cell cycle shift from G0/G1 to G2/M phases, which are known to inhibit progenitor cell differentiation. CFL2 knockdown markedly downregulated the protein expressions of myogenic transcription factors (MyoD, MyoG, and MEF2C) and thereby impaired the differentiation and myotube formation of C2C12 myoblasts. Collectively, this study highlights the roles played by CFL2 on cell cycle progression and proliferation and suggests a novel regulatory mechanism of myogenic differentiation mediated by CFL2.
Sigma-Aldrich
MISSION® esiRNA, targeting human CFL2
Sigma-Aldrich
Phalloidin, Fluorescein Isothiocyanate Labeled, sequence Amanita phalloides(synthetic: peptide sequence)