MilliporeSigma
  • Synthesis of useful fragments in drug discovery: 2-Amino-5-tert-butylpyridine and its oxidised analogues.
Synthesis of useful fragments in drug discovery: 2-Amino-5-tert-butylpyridine and its oxidised analogues.
Bioorganic & medicinal chemistry letters (2011-06-15)
Christopher G Thomson, John Reilly, David A Sandham
ABSTRACT
A novel and robust synthesis of the fragment, 2-amino-5-tert-butylpyridine, has been described, which has been shown to have improved physicochemical properties over 4-tert-butylaniline, when considering drug-like properties. The synthesis also yields fragments containing more highly oxidised precursors to the tert-butyl group as intermediates. These fragments can be incorporated into final target molecules, yielding pharmaceutical compounds and their putative CYP-mediated oxidative metabolites, which can aid in elucidation of metabolic clearance processes.
Sigma-Aldrich
4-tert-Butylaniline, 99%
Pricing and availability is not currently available.