MilliporeSigma
  • Diet drives convergent evolution of gut microbiomes in bamboo-eating species.
Diet drives convergent evolution of gut microbiomes in bamboo-eating species.
Science China. Life sciences (2020-07-04)
Guangping Huang, Xiao Wang, Yibo Hu, Qi Wu, Yonggang Nie, Jiuhong Dong, Yun Ding, Li Yan, Fuwen Wei
ABSTRACT
Gut microbiota plays a critical role in host physiology and health. The coevolution between the host and its gut microbes facilitates animal adaptation to its specific ecological niche. Multiple factors such as host diet and phylogeny modulate the structure and function of gut microbiota. However, the relative contribution of each factor in shaping the structure of gut microbiota remains unclear. The giant (Ailuropoda melanoleuca) and red (Ailurus styani) pandas belong to different families of order Carnivora. They have evolved as obligate bamboo-feeders and can be used as a model system for studying the gut microbiome convergent evolution. Here, we compare the structure and function of gut microbiota of the two pandas with their carnivorous relatives using 16S rRNA and metagenome sequencing. We found that both panda species share more similarities in their gut microbiota structure with each other than each species shares with its carnivorous relatives. This indicates that the specialized herbivorous diet rather than host phylogeny is the dominant driver of gut microbiome convergence within Arctoidea. Metagenomic analysis revealed that the symbiotic gut microbiota of both pandas possesses a high level of starch and sucrose metabolism and vitamin B12 biosynthesis. These findings suggest a diet-driven convergence of gut microbiomes and provide new insight into host-microbiota coevolution of these endangered species.
Sigma-Aldrich
Lysozyme from chicken egg white, powder (crystalline), 70000-140000 U/mg