MilliporeSigma
  • Quantitative Analysis of Pyrazines and Their Perceptual Interactions in Soy Sauce Aroma Type Baijiu.
Quantitative Analysis of Pyrazines and Their Perceptual Interactions in Soy Sauce Aroma Type Baijiu.
Foods (Basel, Switzerland) (2021-03-07)
Yan Yan, Shuang Chen, Yao Nie, Yan Xu
ABSTRACT
Pyrazines are important compounds in soy sauce aroma type Baijiu (SSAB). In this work, a total of 16 pyrazines were analyzed using ultra-performance liquid chromatography coupled with tandem mass spectrometry (UPLC-MS/MS) in SSAB. The quantitative results showed that 2,3,5,6-tetramethylpyrazine, 2,6-dimethylpyrazine and 2,3,5-trimethylpyrazine were the three most concentrated pyrazines. The highest odor activity value (OAV) was determined for 2-ethyl-3,5-dimethylpyrazine. Quantitative analysis combined with descriptive sensory analysis revealed that sub-threshold pyrazines (2,3-dimethylpyrazine, 2,3-diethylpyrazine, 2,3-diethyl-5-methylpyrazine and 2-acetyl-3-methylpyrazine) are significantly correlated with the roasted aroma in SSAB. Our study focused on the impact of sub-threshold pyrazines on the perception of roasted aroma in SSAB. The effect of the sub-threshold pyrazines was detected by the addition of various pyrazines in SSAB samples, despite their sub-threshold concentrations. Furthermore, the presence of sub-threshold pyrazines in dilute alcohol solution resulted in a significant reduction in the odor thresholds of supra-threshold pyrazines. Sensory investigation indicated that pyrazines have a synergistic effect on the perception of roasted aroma. The results highlighted the contribution of some pyrazines to the roasted aroma in SSAB despite their sub-threshold concentrations.