MilliporeSigma
  • Gas sensors based on mass-sensitive transducers. Part 2: Improving the sensors towards practical application.
Gas sensors based on mass-sensitive transducers. Part 2: Improving the sensors towards practical application.
Analytical and bioanalytical chemistry (2020-08-02)
Alexandru Oprea, Udo Weimar
ABSTRACT
Within the framework outlined in the first part of the review, the second part addresses attempts to increase receptor material performance through the use of sensor systems and chemometric methods, in conjunction with receptor preparation methods and sensor-specific tasks. Conclusions are then drawn, and development perspectives for gravimetric sensors are discussed.
Sigma-Aldrich
3-Hexylthiophene, ≥99%