MilliporeSigma
  • Quantifying fluorescent glycan uptake to elucidate strain-level variability in foraging behaviors of rumen bacteria.
Quantifying fluorescent glycan uptake to elucidate strain-level variability in foraging behaviors of rumen bacteria.
Microbiome (2021-01-24)
Leeann Klassen, Greta Reintjes, Jeffrey P Tingley, Darryl R Jones, Jan-Hendrik Hehemann, Adam D Smith, Timothy D Schwinghamer, Carol Arnosti, Long Jin, Trevor W Alexander, Carolyn Amundsen, Dallas Thomas, Rudolf Amann, Tim A McAllister, D Wade Abbott
ABSTRACT
Gut microbiomes, such as the microbial community that colonizes the rumen, have vast catabolic potential and play a vital role in host health and nutrition. By expanding our understanding of metabolic pathways in these ecosystems, we will garner foundational information for manipulating microbiome structure and function to influence host physiology. Currently, our knowledge of metabolic pathways relies heavily on inferences derived from metagenomics or culturing bacteria in vitro. However, novel approaches targeting specific cell physiologies can illuminate the functional potential encoded within microbial (meta)genomes to provide accurate assessments of metabolic abilities. Using fluorescently labeled polysaccharides, we visualized carbohydrate metabolism performed by single bacterial cells in a complex rumen sample, enabling a rapid assessment of their metabolic phenotype. Specifically, we identified bovine-adapted strains of Bacteroides thetaiotaomicron that metabolized yeast mannan in the rumen microbiome ex vivo and discerned the mechanistic differences between two distinct carbohydrate foraging behaviors, referred to as "medium grower" and "high grower." Using comparative whole-genome sequencing, RNA-seq, and carbohydrate-active enzyme fingerprinting, we could elucidate the strain-level variability in carbohydrate utilization systems of the two foraging behaviors to help predict individual strategies of nutrient acquisition. Here, we present a multi-faceted study using complimentary next-generation physiology and "omics" approaches to characterize microbial adaptation to a prebiotic in the rumen ecosystem. Video abstract.
Sigma-Aldrich
Mannan from Saccharomyces cerevisiae, prepared by alkaline extraction
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
CellCrown inserts, 6 well plate inserts with 1.0 μm polycarbonate filter, sterile