MilliporeSigma
  • Determination of Nitrofuran Metabolites in Fish by Ultraperformance Liquid Chromatography-Photodiode Array Detection with Thermostatic Ultrasound-Assisted Derivatization.
Determination of Nitrofuran Metabolites in Fish by Ultraperformance Liquid Chromatography-Photodiode Array Detection with Thermostatic Ultrasound-Assisted Derivatization.
ACS omega (2020-08-11)
Kangkang Wang, Yuli Kou, Meng Wang, Xin Ma, Jide Wang
ABSTRACT
Nitrofuran (NF) is a class of broad-spectrum antibiotics that are used illegally in animal feeding. NF and its metabolites have proven to pose potential risk to human health. To address the current analytical needs to quantify low levels of NF metabolites in animal foods, a sensitive method was developed for simultaneous detection of four NF metabolites in fish products by an ultraperformance liquid chromatography-diode array detector (UPLC-DAD). With 2-nitrobenzaldehyde (2-NBA) as the derivatizing reagent, the metabolites were hydrolyzed and derivatized under the assistance of thermostatic ultrasound. Compared with the current detection methods, the time of the derivatization reaction has been shortened from 16 to 2 h. The relative coefficient of four NF metabolite derivatives reached more than 0.998, with excellent linear relationship. The limits of detection (LODs) and limits of quantification (LOQs) of six repeated determinations reached 0.25-0.33 and 0.80-1.10 μg/kg, respectively. For all four NF metabolites, the limit of detection of the method was below the minimum required performance limit (MRPL) of 1.0 μg/kg, which makes it compatible with the EU requirements. The recoveries ranging from 89.8 to 101.9% with relative standard deviation below 6.5% were obtained for all of the NF metabolites. What's more, this method was successfully applied for the determination of four NF metabolites in the fish products. As a promising approach, this method could also be extended for the quantitation of NF metabolites in aquaculture and poultry products.
Product Description
Supelco
AOZ, VETRANAL®, analytical standard
Sigma-Aldrich
Semicarbazide, 6 wt. % (on silica gel)