MilliporeSigma
  • Microbiota-modulated CART+ enteric neurons autonomously regulate blood glucose.
Microbiota-modulated CART+ enteric neurons autonomously regulate blood glucose.
Science (New York, N.Y.) (2020-08-29)
Paul A Muller, Fanny Matheis, Marc Schneeberger, Zachary Kerner, Veronica Jové, Daniel Mucida
ABSTRACT
The gut microbiota affects tissue physiology, metabolism, and function of both the immune and nervous systems. We found that intrinsic enteric-associated neurons (iEANs) in mice are functionally adapted to the intestinal segment they occupy; ileal and colonic neurons are more responsive to microbial colonization than duodenal neurons. Specifically, a microbially responsive subset of viscerofugal CART+ neurons, enriched in the ileum and colon, modulated feeding and glucose metabolism. These CART+ neurons send axons to the prevertebral ganglia and are polysynaptically connected to the liver and pancreas. Microbiota depletion led to NLRP6- and caspase 11-dependent loss of CART+ neurons and impaired glucose regulation. Hence, iEAN subsets appear to be capable of regulating blood glucose levels independently from the central nervous system.
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Neomycin trisulfate salt hydrate, powder, BioReagent, suitable for cell culture
Supelco
Metronidazole, analytical standard
Sigma-Aldrich
Carmine, powder, ≥42.0% (UV-vis)
Sigma-Aldrich
CTB, ≥98% (HPLC)
Sigma-Aldrich
Fast Green FCF, Dye content ≥85 %
Sigma-Aldrich
Vancomycin hydrochloride from Streptomyces orientalis, ≥900 μg per mg (as vancomycin base)
Sigma-Aldrich
Ampicillin sodium salt, powder or crystals, BioReagent, suitable for cell culture
Supelco
Serotonin, analytical standard
Sigma-Aldrich
Exendin Fragment 9-39, ≥95% (HPLC)