MilliporeSigma
  • Subcellular localization of glypican-5 is associated with dynamic motility of the human mesenchymal stem cell line U3DT.
Subcellular localization of glypican-5 is associated with dynamic motility of the human mesenchymal stem cell line U3DT.
PloS one (2021-02-20)
Masao Takeuchi, Kikuko Takeuchi, Tomoyo Takai, Ritsuko Yamaguchi, Tetsushi Furukawa, Ken-Ichi Akagi, Jun K Takeuchi
ABSTRACT
Glypican-5 (GPC5) is a heparan sulfate proteoglycan (HSPG) localized to the plasma membrane. We previously reported that in the human mesenchymal stem cell line UE6E7T-3, GPC5 is overexpressed in association with transformation and promotes cell proliferation by acting as a co-receptor for Sonic hedgehog signaling. In this study, we found using immunofluorescence microscopy that in transformed cells (U3DT), GPC5 localized not only at primary cilia on the cell surface, but also at the leading edge of migrating cells, at the intercellular bridge and blebs during cytokinesis, and in extracellular vesicles. In each subcellular region, GPC5 colocalized with fibroblast growth factor receptor (FGFR) and the small GTPases Rab11 and ARF6, indicating that GPC5 is delivered to these regions by Rab11-associated recycling endosomes. These colocalizations suggest that GPC5 plays an important role in FGF2 stimulation of cell migration, which was abrogated by knockdown of GPC5. Our findings indicate that GPC5 plays a role in regulation of U3DT cell migration and provides several insights into the functions of GPC5 that could be elucidated by future studies.
Sigma-Aldrich
MISSION® esiRNA, targeting human GPC5