MilliporeSigma
  • Development of a GC-MS Method for Determination of Various Polycyclic Aromatic Hydrocarbons in Iranian Traditional and Semi-industrial Taftoon Bread.
Development of a GC-MS Method for Determination of Various Polycyclic Aromatic Hydrocarbons in Iranian Traditional and Semi-industrial Taftoon Bread.
Iranian journal of pharmaceutical research : IJPR (2021-03-09)
Vahideh Moradi, Seyed Mahdi Seyedain Ardabili, Attaollah Shakoori, Seyed Ebrahim Hoseyni
ABSTRACT
This research reports a validated multi-residue method based on gas chromatography coupled mass spectrometry technique for analysis of 24 Polycyclic Aromatic Hydrocarbons (PAHs) in traditional and semi-industrial Taftoon bread using QuEChERS sample preparation. Matrix effect studies were performed by comparing the slopes of solvent based standard calibration curves and spiked calibration curves. Due to enhancement or suppression effects of matrix, validation of the method was performed using spiked calibration curves. In the concentration range of 10-500 ng/g, the calibration curves for each analyte was linear with a determination coefficient (R2) of 0.991-0.999. The Detection and quantitation limits for the studied PAHs were calculated 0.14-1.49 ng/g and 0.46-4.91 ng/g. The average recoveries for three spiked levels (25, 50 and 200 ng/g), were in the range 77-103% (n = 27), with a satisfactory precision (RSD < 20%). Analysis of Taftoon bread samples using the validated method showed that three compounds; NPH, PHE and ANT were found in 37 (35.2%) samples and in the term of traditional and semi- industrial samples the occurrence of mentioned PAHs were 36.1% and 33.3%, respectively. According to the findings, we proposed that direct flame exposure in gas oven during baking of Taftoon bread could produce PAHs in bread samples.
Sigma-Aldrich
Benzo[k]fluoranthene, for fluorescence, ≥99%
Sigma-Aldrich
meso-Tetraphenylporphyrin, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)
Dibenzo[a,e]pyrene, BCR®, certified reference material
Sigma-Aldrich
Benz[a]anthracene, 99%