MilliporeSigma
  • Pullulan derivative with cationic and hydrophobic moieties as an appropriate macromolecule in the synthesis of nanoparticles for drug delivery.
Pullulan derivative with cationic and hydrophobic moieties as an appropriate macromolecule in the synthesis of nanoparticles for drug delivery.
International journal of biological macromolecules (2020-09-19)
Marieta Constantin, Sanda Bucatariu, Liviu Sacarescu, Oana Maria Daraba, Maria Anghelache, Gheorghe Fundueanu
ABSTRACT
A new amphiphilic pullulan derivative (DBAP-PO) was obtained by grafting tertiary butyl amine and octanoyl groups on the pullulan backbone as cationic and hydrophobic moieties, respectively. The structural characteristics of the modified polymer were investigated by FT-IR and 1H and 13C NMR spectroscopy. The self-association ability in aqueous solution of DBAP-PO was studied by viscosity and fluorescence methods. The intrinsic viscosity of the polymer was determined by Wolf model. The critical aggregation concentration (CAC) value of 0.028 g/dL, determined by fluorescence measurements in the presence of pyrene, was confirmed by capillary viscosimetry and dynamic laser scattering (DLS). Dialysis method was used to demonstrate the capacity of the pullulan derivative to form spherical nanoparticles (d ~ 200 nm) loaded with model drug, sodium diclofenac (DF) (74% entrapment efficiency). The DF release was sustained and pH-dependent. In vitro cytotoxicity as well as morphological studies conducted on the human skin fibroblasts showed that DBAP-PO/DF nanoparticles do not exhibit cytotoxic effects at the pharmacologically relevant concentration of DF, maintaining the typical morphology of the cells.
Sigma-Aldrich
meso-Tetraphenylporphyrin, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)
Sigma-Aldrich
3-(Dibutylamino)propylamine, 98%