MilliporeSigma
  • 3,5-Bis(benzylidene)-1-[3-(2-hydroxyethylthio)propanoyl]piperidin-4-ones: a novel cluster of potent tumor-selective cytotoxins.
3,5-Bis(benzylidene)-1-[3-(2-hydroxyethylthio)propanoyl]piperidin-4-ones: a novel cluster of potent tumor-selective cytotoxins.
Journal of medicinal chemistry (2011-04-01)
Umashankar Das, Hari N Pati, Hiroshi Sakagami, Ken Hashimoto, Masami Kawase, Jan Balzarini, Erik De Clercq, Jonathan R Dimmock
ABSTRACT
Novel 3,5-bis(benzylidene)-1-[3-(2-hydroxyethylthio)propanoyl]piperidin-4-ones (3a-e) display potent cytotoxicity and a preferential lethality toward various neoplasms compared to some normal cells. The corresponding sulfonic acid analogues 5a-e and an isostere 4 demonstrated substantially lower activity. The leads 3d and 3e possess very high activity against colon cancer and leukemia cell lines, caused DNA fragmentation, and activated caspase-3 in HL-60 cells. The enones 3b-e were well tolerated in a short-term toxicity screen in mice.
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
Melphalan, powder