MilliporeSigma
  • Zenglv Fumai Granule protects cardiomyocytes against hypoxia/reoxygenation-induced apoptosis via inhibiting TRIM28 expression.
Zenglv Fumai Granule protects cardiomyocytes against hypoxia/reoxygenation-induced apoptosis via inhibiting TRIM28 expression.
Molecular medicine reports (2021-01-06)
Xiao-Hua Zhang, Hong-Yu Zhao, Yu Wang, Lin Di, Xin-Yu Liu, Feng Qian, Shu-Rong Liu
ABSTRACT
Myocardial ischemia/reperfusion (MIR) injury, which occurs following acute myocardial infarction, can cause secondary damage to the heart. Tripartite interaction motif (TRIM) proteins, a class of E3 ubiquitin ligases, have been recognized as critical regulators in MIR injury. Zenglv Fumai Granule (ZFG) is a clinical prescription for the treatment of sick sinus syndrome, a disease that is associated with MIR injury. The present study aimed to investigate the effect of ZFG on MIR injury and to determine whether ZFG exerts its effects via regulation of TRIM proteins. In order to establish an in vitro MIR model, human cardiomyocyte cell line AC16 was cultured under hypoxia for 5 h and then under normal conditions for 1 h. Following hypoxia/reoxygenation (H/R) treatment, these cells were cultured with different ZFG concentrations. ZFG notably inhibited H/R-induced cardiomyocyte apoptosis. The expression levels of four TRIM proteins, TRIM7, TRIM14, TRIM22 and TRIM28, were also detected. These four proteins were significantly upregulated in H/R-injured cardiomyocytes, whereas their expression was inhibited following ZFG treatment. Moreover, TRIM28 knockdown inhibited H/R-induced cardiomyocyte apoptosis, whereas TRIM28 overexpression promoted apoptosis and generation of reactive oxygen species (ROS) in cardiomyocytes. However, the effects of TRIM28 overexpression were limited by the action of ROS inhibitor N-acetyl-L-cysteine. In addition, the mRNA and protein levels of antioxidant enzyme glutathione peroxidase (GPX)1 were significantly downregulated in H/R-injured cardiomyocytes. TRIM28 knockdown restored GPX1 protein levels but had no effect on mRNA expression levels. Co-immunoprecipitation and ubiquitination assays demonstrated that TRIM28 negatively regulated GPX1 via ubiquitination. In sum, the present study revealed that ZFG attenuated H/R-induced cardiomyocyte apoptosis by regulating the TRIM28/GPX1/ROS pathway. ZFG and TRIM28 offer potential therapeutic options for the treatment of MIR injury.