MilliporeSigma
  • Cell-cell adhesion and 3D matrix confinement determine jamming transitions in breast cancer invasion.
Cell-cell adhesion and 3D matrix confinement determine jamming transitions in breast cancer invasion.
Nature cell biology (2020-08-26)
Olga Ilina, Pavlo G Gritsenko, Simon Syga, Jürgen Lippoldt, Caterina A M La Porta, Oleksandr Chepizhko, Steffen Grosser, Manon Vullings, Gert-Jan Bakker, Jörn Starruß, Peter Bult, Stefano Zapperi, Josef A Käs, Andreas Deutsch, Peter Friedl
ABSTRACT
Plasticity of cancer invasion and metastasis depends on the ability of cancer cells to switch between collective and single-cell dissemination, controlled by cadherin-mediated cell-cell junctions. In clinical samples, E-cadherin-expressing and -deficient tumours both invade collectively and metastasize equally, implicating additional mechanisms controlling cell-cell cooperation and individualization. Here, using spatially defined organotypic culture, intravital microscopy of mammary tumours in mice and in silico modelling, we identify cell density regulation by three-dimensional tissue boundaries to physically control collective movement irrespective of the composition and stability of cell-cell junctions. Deregulation of adherens junctions by downregulation of E-cadherin and p120-catenin resulted in a transition from coordinated to uncoordinated collective movement along extracellular boundaries, whereas single-cell escape depended on locally free tissue space. These results indicate that cadherins and extracellular matrix confinement cooperate to determine unjamming transitions and stepwise epithelial fluidization towards, ultimately, cell individualization.
Sigma-Aldrich
Anti-α-Catenin antibody produced in rabbit, whole antiserum
Sigma-Aldrich
Anti-CD44 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-Uvomorulin/E-Cadherin antibody produced in rat, clone DECMA-1, ascites fluid, buffered aqueous solution
Sigma-Aldrich
Anti-ZEB2 Antibody, from rabbit, purified by affinity chromatography