MilliporeSigma
  • Mycoplasma bovis and other Mollicutes in replacement dairy heifers from Mycoplasma bovis-infected and uninfected herds: A 2-year longitudinal study.
Mycoplasma bovis and other Mollicutes in replacement dairy heifers from Mycoplasma bovis-infected and uninfected herds: A 2-year longitudinal study.
Journal of dairy science (2020-09-29)
M S Hazelton, J M Morton, A M Parker, K L Bosward, P A Sheehy, C J Dwyer, P G Niven, J K House
ABSTRACT
Replacement dairy heifers exposed to Mycoplasma bovis as calves may be at risk of future clinical disease and pathogen transmission, both within and between herds; however, little information is available about these risks. We conducted a 2-yr longitudinal (panel) study starting with 450 heifer calves reared to weaning in 8 herds (7 M. bovis infected with clinical disease, 1 uninfected) under the same ownership. After weaning, heifers were commingled and managed with non-study heifers at a single heifer rearing facility. Nose, conjunctival, and vaginal swabs were collected along with a blood sample at weaning, prebreeding, precalving, and approximately 1 mo postcalving. Additionally, a colostrum sample was collected upon calving and a composite milk sample was collected 1 mo postcalving. The swabs, colostrum, and milk samples were cultured for Mycoplasma spp., and serum from the blood was evaluated for serological evidence of exposure to M. bovis using an ELISA. Despite a high M. bovis ELISA seroprevalence at weaning in the heifers from the 7 M. bovis-infected herds with clinical disease [72% (289/400); range by herd: 28-98%], M. bovis was isolated from only 4% (16/400) of the same heifers at the same time. In heifers from the uninfected herd at weaning, M. bovis seroprevalence was 2% (1/50) and M. bovis was not detected by culture. Mycoplasma bovis was isolated from 0.5% (2/414) of heifers at prebreeding, 0% (0/374) of heifers at precalving, and 0.3% (1/356) of heifers 1 mo postcalving. The nose was the predominant anatomical site of M. bovis colonization (74%; 14/19 culture positives). A single heifer (from an M. bovis-infected herd with clinical disease) was repeatedly detected with M. bovis in its nose at weaning, prebreeding, and postcalving samplings. This demonstrates the possibility, albeit rare, of a long-term M. bovis carrier state in replacement heifers exposed to M. bovis as calves, up to at least 1 mo after entry into the milking herd. No M. bovis clinical disease was detected in any heifer from weaning through to the end of the study (approximately 1 mo after calving). Acholeplasma spp. were commonly isolated throughout the study. Mycoplasma bovigenitalium, Mycoplasma bovoculi, and Mycoplasma bovirhinis were isolated infrequently. Mycoplasma bovis seroprevalences at prebreeding, precalving, and postcalving samplings were 27% (112/414), 12% (46/374), and 18% (65/356), respectively. Overall, the results show that replacement heifers from groups exposed to M. bovis preweaning can become colonized with M. bovis and that colonization can, uncommonly, be present after their first calving. For groups of 50 or more heifers exposed to M. bovis preweaning, there is at least a nontrivial probability that the group will contain at least 1 shedding heifer postcalving.
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from calf thymus, Type I, fibers