MilliporeSigma
  • Peripheral serotonin regulates glucose and insulin metabolism in Holstein dairy calves.
Peripheral serotonin regulates glucose and insulin metabolism in Holstein dairy calves.
Domestic animal endocrinology (2020-08-03)
S L Field, M G Marrero, B Dado-Senn, A L Skibiel, P M Ramos, T L Scheffler, J Laporta
ABSTRACT
Peripheral serotonin regulates energy metabolism in several mammalian species, however, the potential contribution of serotonergic mechanisms as metabolic and endocrine regulators in growing dairy calves remain unexplored. Objectives were to characterize the role of serotonin in glucose and insulin metabolism in dairy calves with increased serotonin bioavailability. Milk replacer was supplemented with saline, 5-hydroxytryptophan (90 mg/d), or fluoxetine (40 mg/d) for 10-d (n = 8/treatment). Blood was collected daily during supplementation and on days 2, 7, and 14 during withdrawal. Calves were euthanized after 10-d supplementation or 14-d withdrawal periods to harvest liver and pancreas tissue. 5-hydroxytryptophan increased circulating insulin concentrations during the supplementation period, whereas both treatments increased circulating glucose concentration during the withdrawal period. The liver and pancreas of preweaned calves express serotonin factors (ie, TPH1, SERT, and cell surface receptors), indicating their ability to synthesize, uptake, and respond to serotonin. Supplementation of 5-hydroxytryptophan increased hepatic and pancreatic serotonin concentrations. After the withdrawal period, fluoxetine cleared from the pancreas but not liver tissue. Supplementation of 5-hydroxytryptophan upregulated hepatic mRNA expression of serotonin receptors (ie, 5-HTR1B, -1D, -2A, and -2B), and downregulated pancreatic 5-HTR1F mRNA and insulin-related proteins (ie, Akt and pAkt). Fluoxetine-supplemented calves had fewer pancreatic islets per microscopic field with reduced insulin intensity, whereas 5-hydroxytryptophan supplemented calves had increased islet number and area with greater insulin and serotonin and less glucagon intensities. After the 14-d withdrawal of 5-hydroxytryptophan, hepatic mRNA expression of glycolytic and gluconeogenic enzymes were simultaneously downregulated. Improving serotonin bioavailability could serve as a potent regulator of endocrine and metabolic processes in dairy calves.
Product Description
Supelco
Serotonin, analytical standard
Sigma-Aldrich
5-Hydroxy-L-tryptophan, powder