MilliporeSigma
  • Physiological Concentration of H2O2 Supports Dopamine Neuronal Survival via Activation of Nrf2 Signaling in Glial Cells.
Physiological Concentration of H2O2 Supports Dopamine Neuronal Survival via Activation of Nrf2 Signaling in Glial Cells.
Cellular and molecular neurobiology (2020-04-23)
Guoqing Wang, Qiuyu Yang, Changqing Zheng, Daidi Li, Jingjie Li, Feng Zhang
ABSTRACT
Traditionally, hydrogen peroxide (H2O2) was formed from cellular oxidative metabolism and often viewed as toxic waste. In fact, H2O2 was a benefit messenger for neuron-glia signaling and synaptic transmission. Thus, H2O2 was a double-edged sword and neuroprotection vs. neurotoxicity produced by H2O2 was difficult to define. Nuclear factor erythroid 2-related factor 2 (Nrf2) has been implicated as an intracellular regulator of neuronal growth. Inactivation of Nrf2 participated in the development of Parkinson's disease (PD). Thus, suitable activation of Nrf2 was essential for the prevention and treatment of PD. This study aimed to explore whether H2O2-conferred neuroprotective effects to support neuronal survival. H2O2 were added into primary neuron-glia, neuron-astroglia and neuron-microglia co-cultures in concentration- and time-dependent manners. H2O2 increased dopamine (DA) neuronal survival in concentration- and time-dependent manners. In addition, glial cells Nrf2 activation involved in H2O2-supported DA neuronal survival with the following phenomenons. First, H2O2 activated Nrf2 signaling pathway. Second, H2O2 generated beneficial neuroprotection in neuron-glia, neuron-astroglia and neuron-microglia co-cultures but not in neuron-enriched cultures. Third, silence of Nrf2 in glial cells abolished H2O2-conferred DA neuronal survival. This study demonstrated that physiological concentration of H2O2-supported DA neuronal survival via activation of Nrf2 signaling in glial cells. Our data permit to re-evaluate the role of H2O2 in the pathogenesis and therapeutic strategies for PD.
Sigma-Aldrich
Leu-Leu