MilliporeSigma
  • The ABC transporter AnrAB contributes to the innate resistance of Listeria monocytogenes to nisin, bacitracin, and various beta-lactam antibiotics.
The ABC transporter AnrAB contributes to the innate resistance of Listeria monocytogenes to nisin, bacitracin, and various beta-lactam antibiotics.
Antimicrobial agents and chemotherapy (2010-07-21)
Barry Collins, Nicola Curtis, Paul D Cotter, Colin Hill, R Paul Ross
ABSTRACT
A mariner transposon bank was used to identify loci that contribute to the innate resistance of Listeria monocytogenes to the lantibiotic nisin. In addition to highlighting the importance of a number of loci previously associated with nisin resistance (mprF, virRS, and telA), a nisin-sensitive phenotype was associated with the disruption of anrB (lmo2115), a gene encoding the permease component of an ABC transporter. The contribution of anrB to nisin resistance was confirmed by the creation of nonpolar deletion mutants. The loss of this putative multidrug resistance transporter also greatly enhanced sensitivity to bacitracin, gallidermin, and a selection of β-lactam antibiotics. A comparison of the relative antimicrobial sensitivities of a number of mutants established the ΔanrB strain as being one of the most bacitracin-sensitive L. monocytogenes strains identified to date.
Sigma-Aldrich
Carbenicillin, Ready Made Solution, 100 mg/mL in ethanol/water, 0.2 μm filtered
Sigma-Aldrich
Cefaclor
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)