MilliporeSigma
  • Nanomechanical Insights into Versatile Polydopamine Wet Adhesive Interacting with Liquid-Infused and Solid Slippery Surfaces.
Nanomechanical Insights into Versatile Polydopamine Wet Adhesive Interacting with Liquid-Infused and Solid Slippery Surfaces.
ACS applied materials & interfaces (2021-02-02)
Lei Xie, Xin Cui, Jing Liu, Qiuyi Lu, Jun Huang, Xiaohui Mao, Diling Yang, Jinglin Tan, Hao Zhang, Hongbo Zeng
ABSTRACT
Mussel-inspired polydopamine (PDA) can be readily deposited on almost all kinds of substrates and possesses versatile wet adhesion. Meanwhile, slippery surfaces have attracted much attention for their self-cleaning capabilities. It remains unclear how the versatile PDA adhesive would interact with slippery surfaces. In this work, both liquid-infused poly(tetrafluoroethylene) (PTFE) (LI-PTFE) and solid slippery surfaces (i.e., self-assembly of small thiol-terminated organosilane, polysiloxane covalently attached to substrates) were fabricated to investigate their capability to prevent PDA deposition. It was found that PDA particles could be easily deposited on a PTFE membrane and the two types of solid slippery surfaces, which resulted in the alternation of their surface wettability and slippery behavior of water droplets. Adhesion was detected between a PDA-coated silica colloidal probe and the PTFE membrane or solid slippery surfaces through quantitative force measurements using an atomic force microscope (AFM), mainly due to van der Waals (vdW) and hydrophobic interactions, which led to the PDA deposition phenomenon. In contrast, LI-PTFE with a thin liquid lubricant film could effectively prevent PDA deposition, with negligible changes in surface morphology, wettability, and slippery characteristics. Although PDA particles could be loosely attached to the lubricant/water interface for LI-PTFE based on the capillary adhesion measured by AFM, they could be readily removed by gentle rinsing with water, as demonstrated by the ultralow friction over LI-PTFE as compared to PTFE using lateral force microscopy (LFM). Our results indicate that LI-PTFE possesses excellent antifouling and self-cleaning properties even when interacting with the versatile PDA wet adhesives. This work provides new insights into the deposition of PDA on slippery surfaces and their interaction mechanism at the nanoscale, with useful implications for the design and development of novel slippery surfaces.
Sigma-Aldrich
Dimethoxydimethylsilane, 95%
Sigma-Aldrich
(3-Mercaptopropyl)methyldimethoxysilane, ≥95.0%