MilliporeSigma
  • The synergistic tumor growth-inhibitory effect of probiotic Lactobacillus on transgenic mouse model of pancreatic cancer treated with gemcitabine.
The synergistic tumor growth-inhibitory effect of probiotic Lactobacillus on transgenic mouse model of pancreatic cancer treated with gemcitabine.
Scientific reports (2020-11-25)
Shan-Ming Chen, Wee-Wei Chieng, Szu-Wei Huang, Li-Jin Hsu, Ming-Shiou Jan
ABSTRACT
Pancreatic cancer is one of the most lethal and chemo-resistant cancers worldwide. Growing evidence supports the theory that the gut microbiota plays an essential role in modulating the host response to anti-cancer therapy. The present study aimed to explore the effect of probiotics as an adjuvant during chemotherapy for pancreatic cancer. An LSL-KrasG12D/--Pdx-1-Cre mouse model of pancreatic ductal adenocarcinoma (PDAC) was created to study the effects of using four-week multi-strain probiotics (Lactobacillus paracasei GMNL-133 and Lactobacillus reuteri GMNL-89) as an adjuvant therapy for controlling cancer progression. At 12 weeks of age, pancreatitis was induced in the mice by two intraperitoneal injection with caerulein (25 μg/kg 2 days apart). Over the next 4 weeks the mice were treated with intraperitoneal injections of gemcitabine in combination with the oral administration of probiotics. The pancreas was then harvested for analysis. Following caerulein treatment, the pancreases of the LSL-KrasG12D/--Pdx-1-Cre transgenic mice exhibited more extensive pancreatic intraepithelial neoplasia (PanIN) formation. Combined treatment with gemcitabine and probiotics revealed a lower grade of PanIN formation and a decrease in the expression of vimentin and Ki-67. Mice that received gemcitabine in combination with probiotics had lower aspartate aminotransferase (AST) and alanine aminotransferase (ALT) levels. Notably, the use of high-dose probiotics alone without gemcitabine also had an inhibitory effect on PanIN changes and serum liver enzyme elevation. These findings suggest that probiotics are able to make standard chemotherapy more effective and could help improve the patient's tolerance of chemotherapy.
Sigma-Aldrich
Caerulein, ≥95% (HPLC)