MilliporeSigma
  • Dual drug delivery from hydrophobic and hydrophilic intraocular lenses: in-vitro and in-vivo studies.
Dual drug delivery from hydrophobic and hydrophilic intraocular lenses: in-vitro and in-vivo studies.
Journal of controlled release : official journal of the Controlled Release Society (2020-07-20)
Ana Topete, Junmei Tang, Xiaoxu Ding, Helena P Filipe, Jorge A Saraiva, Ana Paula Serro, Quankui Lin, Benilde Saramago
ABSTRACT
Posterior capsule opacification (PCO) still remains the most frequent long term complication after cataract surgery, while endophthalmitis is rare but severe and should be prevented at all cost. Intraocular lenses (IOLs) with different designs (eg. edge and body-haptics angle) and materials (acrylic hydrophobic and acrylic hydrophilic surfaces) have been studied to reduce PCO. For the prevention of endophthalmitis, intracameral injection followed or not by topical treatment with antibiotics and anti-inflammatories are usually prescribed. The objective of this work was to investigate the use of IOLs as controlled release platforms of two drugs, the antibiotic moxifloxacin (MXF) and the anti-inflammatory ketorolac (KTL) that could advantageously substitute the usual treatment. Two types of IOLs were chosen, hydrophobic and hydrophilic. Hydrophobic IOLs have shown better results in the prevention of PCO because they adhere better to the posterior capsular bag, while hydrophilic IOLs are advised in the case of patients with uveitis, glaucoma or diabetes. The IOLs were loaded with MXF + KTL and sterilized by high hydrostatic pressure. Both IOLs reduced the tendency for adhesion of LECs. In vivo tests were done to compare the concentration of the drugs in the aqueous humor obtained after eye drops administration and drug-loaded IOLs implantation. The developed IOLs were able to release MXF and KTL at therapeutic levels, in a sustained way, which contrasts with the eye drops prophylaxis. No PCO signs were detected and histological analyses demonstrated biocompatibility of these devices.
Sigma-Aldrich
Moxifloxacin hydrochloride, ≥98% (HPLC)