MilliporeSigma
  • Zinc supplementation alleviates endoplasmic reticulum stress during porcine oocyte in vitro maturation by upregulating zinc transporters.
Zinc supplementation alleviates endoplasmic reticulum stress during porcine oocyte in vitro maturation by upregulating zinc transporters.
Journal of cellular physiology (2020-09-19)
Muhammad Rosyid Ridlo, Geon A Kim, Anukul Taweechaipaisankul, Eui Hyun Kim, Byeong Chun Lee
ABSTRACT
Endoplasmic reticulum (ER) stress is a major contributor to embryonic development failure. Mammalian oocytes have a high risk of exposure to cellular stress during in vitro embryo production. We investigated the effects of zinc supplementation during in vitro maturation under ER stress. We evaluated cumulus expansion, embryonic development derived by parthenogenetic activation, reactive oxygen species, protein expression of X-box binding protein 1 (XBP1), and expression of genes related to ER stress. Supplementation with 1 μg/ml zinc significantly increased the nuclear maturation of oocytes, cleavage and blastocyst formation rates, and total blastocyst cell number (p < .05). Under ER stress, zinc significantly reduced protein expression of XBP1, and increased cleavage and blastocyst rates (p < .05). Concomitantly, zinc supplementation upregulated the expression of zinc transporters (SLC39A14 and SLC39A10), PTGS2, and downregulated ER stress-related genes (sXBP1, uXBP1, ATF4, and PTPN1/PTP1B), and caspase 3. These results suggest that zinc supplementation alleviates ER stress by providing essential metal-ion transporters for oocyte maturation and subsequent embryonic development.
Sigma-Aldrich
Zinc sulfate heptahydrate, BioUltra, 2.0 M in H2O