MilliporeSigma
  • Inhalation of odors containing DMHF generated by the Maillard reaction affects physiological parameters in rats.
Inhalation of odors containing DMHF generated by the Maillard reaction affects physiological parameters in rats.
Scientific reports (2020-08-20)
Issei Yokoyama, Motoko Ohata, Yusuke Komiya, Jun Nagasao, Keizo Arihara
ABSTRACT
The effects of odors generated by the Maillard reaction from amino acids and reducing sugars on physiological parameters (blood pressure, heart rate, and oxidative stress levels) in Wistar rats were investigated in the present study. The Maillard reaction samples were obtained from glycine, arginine, or lysine of 1.0 mol/L and glucose of 1.0 mol/L with heat treatment. The odor-active compounds in the Maillard reaction samples were identified using the aroma extract dilution analysis. Among the odor-active compounds identified, 2,5-dimethyl-4-hydroxy-3(2H)-furanone (DMHF, FURANEOL and strawberry furanone) had the highest odor activity and its concentration was affected by amino acid types. The Maillard reaction odors generated from glycine or arginine significantly decreased systolic blood pressure and heart rate in rats when inhaled. These physiological effects were associated with DMHF. Furthermore, oxidative stress marker levels in rat plasma were decreased by the inhalation of DMHF. The inhalation of DMHF appears to at least partly affect physiological parameters by decreasing oxidative stress.
Sigma-Aldrich
2,3-Dimethylpyrazine, ≥95%, FCC, FG