MilliporeSigma
  • Molecular characterization, immune and xenobiotic responses of glutathione S-transferase omega 1 from the big-belly seahorse: Novel insights into antiviral defense.
Molecular characterization, immune and xenobiotic responses of glutathione S-transferase omega 1 from the big-belly seahorse: Novel insights into antiviral defense.
Fish & shellfish immunology (2020-12-22)
H M V Udayantha, D S Liyanage, Kishanthini Nadarajapillai, W K M Omeka, Hyerim Yang, Taehyug Jeong, Jehee Lee
ABSTRACT
Glutathione S-transferases (GSTs) are important enzymes involved in phase II detoxification and function by conjugating with the thiol group of glutathione. In this study, we isolated an omega class GST from the big-belly seahorse (Hippocampus abdominalis; HaGSTO1) to study the putative xenobiotic responses and defense ability against viral and bacterial infections in this animal. The isolated HaGSTO1 gene, with a cording sequence of 720 bp, encodes a peptide of 239 amino acids. The predicted molecular mass and theoretical isoelectric point of HaGSTO1 was 27.47 kDa and 8.13, respectively. In-silico analysis of HaGSTO1 revealed a characteristic N-terminal thioredoxin-like domain and a C-terminal domain. Unlike other GSTs, the C-terminal of HaGSTO1 reached up to the N-terminal, and the N-terminal functional group was cysteine rather than tyrosine or serine, as observed in other GSTs. Phylogenetic analysis showed the evolutionary proximity of HaGSTO1 with other identified vertebrate and invertebrate GST orthologs. For the first time, we demonstrated the viral defense capability of HaGSTO1 against viral hemorrhagic septicemia virus (VHSV) infection. All six nucleoproteins of VHSV were significantly downregulated in HaGSTO1-overexpressing FHM cells at 24 h after infection compared with those in the control. Moreover, arsenic toxicity was significantly reduced in HaGSTO1-overexpressing FHM cells, and cell viability increased. Real-time polymerase chain reaction analysis showed that HaGSTO1 transcripts were highly expressed in the pouch and gill when compared with those in other tissues. Blood HaGSTO1 transcripts were significantly upregulated after Edwardsiella tarda, Streptococcus iniae, lipopolysaccharide, and polyinosinic:polycytidylic acid challenge experiments. Collectively, these findings suggest the involvement of HaGSTO1 in the host defense mechanism of seahorses.
Sigma-Aldrich
4-Nitrobenzyl chloride, 99%