MilliporeSigma
  • A Novel Enzyme-Based SPR Strategy for Detection of the Antimicrobial Agent Chlorophene.
A Novel Enzyme-Based SPR Strategy for Detection of the Antimicrobial Agent Chlorophene.
Biosensors (2021-02-13)
Gabriela Elizabeth Quintanilla-Villanueva, Donato Luna-Moreno, Edgar Allan Blanco-Gámez, José Manuel Rodríguez-Delgado, Juan Francisco Villarreal-Chiu, Melissa Marlene Rodríguez-Delgado
ABSTRACT
Chlorophene is an important antimicrobial agent present in disinfectant products which has been related to health and environmental effects, and its detection has been limited to chromatographic techniques. Thus, there is a lack of research that attempts to develop new analytical tools, such as biosensors, that address the detection of this emerging pollutant. Therefore, a new biosensor for the direct detection of chlorophene in real water is presented, based on surface plasmon resonance (SPR) and using a laccase enzyme as a recognition element. The biosensor chip was obtained by covalent immobilization of the laccase on a gold-coated surface through carbodiimide esters. The analytical parameters accomplished resulted in a limit of detection and quantification of 0.33 mg/L and 1.10 mg/L, respectively, fulfilling the concentrations that have already been detected in environmental samples. During the natural river's measurements, no significant matrix effects were observed, obtaining a recovery percentage of 109.21% ± 7.08, which suggested that the method was suitable for the fast and straightforward analysis of this contaminant. Finally, the SPR measurements were validated with an HPLC method, which demonstrated no significant difference in terms of precision and accuracy, leading to the conclusion that the biosensor reflects its potential as an alternative analytical tool for the monitoring of chlorophene in aquatic environments.
Sigma-Aldrich
2-Benzyl-4-chlorophenol, 95%