MilliporeSigma
  • Nutritional quality of Drosophila melanogaster as factitious prey for rearing the predatory bug Orius majusculus.
Nutritional quality of Drosophila melanogaster as factitious prey for rearing the predatory bug Orius majusculus.
Insect science (2020-01-29)
Marta Montoro, Henrik H De Fine Licht, Lene Sigsgaard
ABSTRACT
The predatory bug, Orius majusculus (Reuter), is an important predator of thrips commercially produced for augmentative releases using the eggs of the Mediterranean flour moth Ephestia kuehniella (Zeller). In this study, we assessed the potential for using frozen adults of fruit flies, Drosophila melanogaster (Meigen), either as nymphal rearing diet or as diet throughout the entire life-cycle. We compared life-history traits and reproduction of predators when fed D. melanogaster with high lipid body content (lipid-rich) and with high protein body content (protein-rich), using a diet of 100% E. kuehniella eggs as control. We also analyzed the biochemical composition of both prey and predator in order to assess the nutritional quality of each diet, which partially explained the adequacy of the different diets for O. majusculus. There were significant differences between predators fed the two types of D. melanogaster, with the protein-rich flies as diet providing the best results in terms of mortality and fecundity. Furthermore, we show that while feeding O. majusculus throughout their development with D. melanogaster increases mortality and reduces reproduction, protein-rich D. melanogaster can be used as nymphal diet with minimal reduction in reproductivity and minimal increase in mortality.
Sigma-Aldrich
Casein from bovine milk, suitable for substrate for protein kinase (after dephosphorylation), purified powder
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)