MilliporeSigma
  • Characterization of 4-methyl-2-oxo-1,2-dihydroquinolin-6-yl acetate as an effective antiplatelet agent.
Characterization of 4-methyl-2-oxo-1,2-dihydroquinolin-6-yl acetate as an effective antiplatelet agent.
Bioorganic & medicinal chemistry (2010-05-08)
Nivedita Priya, Anjali Gupta, Karam Chand, Prabhjot Singh, Abha Kathuria, Hanumantharao G Raj, Virinder S Parmar, Sunil K Sharma
ABSTRACT
We have studied earlier a membrane bound novel enzyme Acetoxy Drug: protein transacetylase identified as Calreticulin Transacetylase (CRTAase) that catalyzes the transfer of acetyl groups from polyphenolic acetates (PAs) to the receptor proteins and thus modulating their biological activities. In this communication, we have reported for the first time that acetoxy quinolones are endowed with antiplatelet action by virtue of causing CRTAase catalyzed activation of platelet Nitric Oxide Synthase (NOS) by way of acetylation leading to the inhibition of ADP/Arachidonic acid (AA)-dependent platelet aggregation. The correlation of specificity of platelet CRTAase to various analogues of acetoxy quinolones with intracellular NO and consequent effect on inhibition of platelet aggregation was considered crucial. Among acetoxy quinolones screened, 6-AQ (4-methyl-2-oxo-1,2-dihydroquinolin-6-yl acetate/6-acetoxyquinolin-2-one, 22) was found to be the superior substrate to platelet CRTAase and emerged as the most active entity to produce antiplatelet action both in vitro and in vivo. 6-AQ caused the inhibition of cyclooxygenase-1 (Cox-1) resulting in the down regulation of thromboxane A2 (TxA2) and the inhibition of platelet aggregation. Structural modification of acetoxy quinolones positively correlated with enhancement of intracellular NO and antiplatelet action.
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Sigma-Aldrich
Aspirin, meets USP testing specifications