MilliporeSigma
  • TIMM29 interacts with hepatitis B virus preS1 to modulate the HBV life cycle.
TIMM29 interacts with hepatitis B virus preS1 to modulate the HBV life cycle.
Microbiology and immunology (2020-09-25)
Nelly Gakii Muriungi, Keiji Ueda
ABSTRACT
Hepatitis B virus (HBV), a major global health problem, can cause chronic hepatitis, liver cirrhosis, and hepatocellular carcinomas in chronically infected patients. However, before HBV infection can be adequately controlled, many mysteries about the HBV life cycle must be solved. In this study, TIMM29, an inner mitochondrial membrane protein, was identified as an interaction partner of the preS1 region of the HBV large S protein. The interaction was verified by both an immunoprecipitation with preS1 peptides and a GST-pulldown assay. Immunofluorescence studies also showed colocalization of preS1 and TIMM29. Moreover, it was determined that the preS1 bound with amino acids 92-189 of the TIMM29 protein. Infection of HBV in TIMM29-overexpressing NTCP/G2 cells resulted in a significant decrease of HBeAg and both extracellular particle-associated and core particle-associated HBV DNA without affecting cccDNA formation. Comparable results were obtained with TIMM29-overexpressing HB611 cells, which constitutively produce HBV. In contrast, knockout of TIMM29 in NTCP/G2 cells led to a higher production of HBV including HBeAg expression, as did knockout of TIMM29 in HB611. Collectively, these results suggested that TIMM29 interacts with the preS1 region of the HBV large S protein and modulates HBV amplification.
Sigma-Aldrich
Anti-β-Tubulin antibody, Mouse monoclonal, clone TUB 2.1, purified from hybridoma cell culture
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
Anti-TIMM29 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-SAMM50 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution