MilliporeSigma
  • Improved water solubility and photodynamic activity of hydroxy-modified porphyrins by complexation with cyclodextrin.
Improved water solubility and photodynamic activity of hydroxy-modified porphyrins by complexation with cyclodextrin.
Organic & biomolecular chemistry (2020-08-22)
Takehiro Yumoto, Shuhei Satake, Shodai Hino, Kouta Sugikawa, Riku Kawasaki, Atsushi Ikeda
ABSTRACT
Porphyrin derivatives with hydroxyphenyl or dihydroxyphenyl moieties in the meso-position were able to dissolve in water by complexation with two trimethyl-β-cyclodextrins (TMe-β-CDxs) without further chemical modification of porphyrins or addition of dimethyl sulfoxide as a co-solvent. TMe-β-CDx-complexed tetra(hydroxyphenyl)porphyrins (THPPs) with phenols in meso-positions have a much higher photodynamic activity than TMe-β-CDx-complexed tetrakis(dihydroxyphenyl)porphyrins (TDHPPs) with dihydroxyphenyl in the meso-position. The main reason for the different photoactivity is due to the intracellular uptakes of these complexes. Thus phenols in meso-positions of porphyrin derivatives in the complexes were recognized by HeLa cell receptors. Furthermore, the photodynamic activity of TMe-β-CDx-complexed THPP was much higher than that of THPP injected as a dimethyl sulfoxide solution.
Sigma-Aldrich
9,10-Anthracenediyl-bis(methylene)dimalonic acid, BioReagent, suitable for fluorescence, ≥90% (HPCE)