MilliporeSigma
  • Surface Laser-Marking and Mechanical Properties of Acrylonitrile-Butadiene-Styrene Copolymer Composites with Organically Modified Montmorillonite.
Surface Laser-Marking and Mechanical Properties of Acrylonitrile-Butadiene-Styrene Copolymer Composites with Organically Modified Montmorillonite.
ACS omega (2020-08-11)
Guangwei Lu, Yinqiu Wu, Yang Zhang, Kailun Wang, Hongxin Gao, Keming Luo, Zheng Cao, Junfeng Cheng, Chunlin Liu, Lei Zhang, Juan Qi
ABSTRACT
In this study, organically modified montmorillonite (OMMT) was prepared by modifying MMT with a cationic surfactant cetyltrimethylammonium bromide (CTAB). The obtained OMMT of different loading contents (1, 2, 4, 6, and 8 wt %) was melt-blended with poly(acrylonitrile-co-butadiene-co-styrene) (ABS) to prepare a series of ABS/OMMT composites, which were laser marked using a neodymium-doped yttrium aluminum garnet (Nd:YAG) laser beam of 1064 nm under different laser current processes. X-ray diffraction (XRD), color difference spectrometer, optical microscope, water contact angle tests, scanning electron microscope (SEM), and Raman spectroscopy were carried out to characterize the morphology, structure, and properties of the laser-patterned ABS composites. The effects of the addition of OMMT and the laser marking process on the mechanical properties of ABS/OMMT composites were investigated through mechanical property tests. The results show that the obtained ABS/OMMT composites have enhanced laser marking performance, compared to the ABS. When the OMMT content is 2 wt % and the laser current intensity is 9 A, the marking on ABS composites has the highest contrast (ΔE = 36.38) and sharpness, and the quick response (QR) code fabricated can be scanned and identified with a mobile app. SEM and water contact angle tests showed that the holes, narrow cracks, and irregular protrusion are formed on the composite surface after laser marking, resulting in a more hydrophobic surface and an increased water contact angle. Raman spectroscopy and XRD indicate that OMMT can absorb the near-infrared laser energy, undergo photo thermal conversion, and cause the pyrolysis and carbonization of ABS to form black marking, and the crystal structure itself does not change significantly. When the 2 wt % of OMMT is loaded, the tensile strength, elongation at break, and impact strength of ABS/OMMT are increased by 15, 20, and 14%, respectively, compared to ABS. Compared with the unmarked ABS/OMMT, the defects including holes and cracks generated on the surface of the marked one lead to the decreased mechanical property. The desirable combination of high contrast laser marking performance and mechanical properties can be achieved at an OMMT loading content of 2 wt % and a laser current intensity of 9 A. This research work provides a simple, economical, and environmentally friendly method for laser marking of engineering materials such as ABS.
Sigma-Aldrich
3-(Trimethoxysilyl)propyl methacrylate, 98%