MilliporeSigma
  • A Combined Experimental and Theoretical Study of Screen-printing High Transparent Conductive Mesoscopic ITO Films.
A Combined Experimental and Theoretical Study of Screen-printing High Transparent Conductive Mesoscopic ITO Films.
Scientific reports (2020-03-21)
Feiyang Qiao, Lei Lu, Pingcheng Han, Dekai Ge, Yunjun Rui, Dawei Gu, Tianyou Zhang, Jiwei Hou, Ying Yang
ABSTRACT
We have successfully fabricated transparent conductive mesoporous indium tin oxide (TCM-ITO) films by a screen-printing method. The TCM-ITO films possess approximately 22 nm mesopores and obtain electrical conductivity up to 14.96 S/cm by adjusting the mass ratio of cubic-shaped ITO nanoparticles to ethyl cellulose (EC) and precisely controlling the annealing process. The regulation mechanism of EC and the heat-induced recrystallization process of ITO nanoparticles are elaborated. The internal kinetic processes of the films based on different surface states are analysed, and an extensible impedance model is established.
Sigma-Aldrich
Tin(IV) chloride pentahydrate, 98%