MilliporeSigma
  • Preweanling administration of terazosin decreases blood pressure of hypertensive rats in adulthood.
Preweanling administration of terazosin decreases blood pressure of hypertensive rats in adulthood.
Hypertension (Dallas, Tex. : 1979) (1996-05-01)
R McCarty, J H Lee
ABSTRACT
To examine the contribution of the sympathetic nervous system to the development of hypertension, we injected spontaneously hypertensive rat (SHR) pups and normotensive Wistar-Kyoto rat (WKY) pups twice daily with saline (1.0 mL/kg SC) or terazosin (0.5 mg/kg SC), an alpha 1-adrenoceptor antagonist, from postnatal day 1 through 21. We determined the effectiveness and duration of action of this terazosin dose in pilot studies with adult SHR and WKY. Body weights of WKY pups were greater than body weights of SHR pups from postnatal day 1 through 21. In addition, body weights of terazosin-treated pups of both strains were comparable to body weights of saline-injected littermate controls. Indirectly measured systolic pressures of terazosin-treated SHR were reduced significantly at 60 and 90 days of age but not at 30 days of age compared with saline-injected littermate controls. Terazosin did not affect systolic pressures of WKY, measured at 30, 60, and 90 days of age. At 100 days of age, in chronically catheterized rats, mean arterial pressures of terazosin-treated SHR were reduced significantly compared with those of saline-injected littermate controls. In contrast, terazosin did not affect mean arterial pressures of WKY at 100 days of age. Finally, preweanling treatment with terazosin did not alter patterns of open field behavior of adult SHR or WKY. SHR were significantly more active and reared more frequently compared with WKY. These findings indicate that the time between birth and weaning at 21 days of age is critical for the full expression of the hypertensive phenotype in SHR. Chronic blockage of alpha 1-adrenoceptors during the preweanling period in SHR may reduce vascular hypertrophy, leading to long-term reductions in arterial pressure.
Sigma-Aldrich
Terazosin hydrochloride, ≥98% (TLC), powder