MilliporeSigma
  • Nox2+ myeloid cells drive vascular inflammation and endothelial dysfunction in heart failure after myocardial infarction via angiotensin II receptor type 1.
Nox2+ myeloid cells drive vascular inflammation and endothelial dysfunction in heart failure after myocardial infarction via angiotensin II receptor type 1.
Cardiovascular research (2020-02-23)
Michael Molitor, Wolf-Stephan Rudi, Venkata Garlapati, Stefanie Finger, Rebecca Schüler, Sabine Kossmann, Jeremy Lagrange, Thanh Son Nguyen, Johannes Wild, Tanja Knopp, Susanne H Karbach, Maike Knorr, Wolfram Ruf, Thomas Münzel, Philip Wenzel
ABSTRACT
Heart failure (HF) ensuing myocardial infarction (MI) is characterized by the initiation of a systemic inflammatory response. We aimed to elucidate the impact of myelomonocytic cells and their activation by angiotensin II on vascular endothelial function in a mouse model of HF after MI. HF was induced in male C57BL/6J mice by permanent ligation of the left anterior descending coronary artery. Compared to sham, HF mice had significantly impaired endothelial function accompanied by enhanced mobilization of Sca-1+c-Kit+ haematopoietic stem cells and Sca-1-c-Kit+ common myeloid and granulocyte-macrophage progenitors in the bone marrow as well as increased vascular infiltration of CD11b+Ly6G-Ly6Chigh monocytes and accumulation of CD11b+ F4/80+ macrophages, assessed by flow cytometry. Using mice with Cre-inducible expression of diphtheria toxin receptor in myeloid cells, we selectively depleted lysozyme M+ myelomonocytic cells for 10 days starting 28 days after MI. While the cardiac phenotype remained unaltered until 38 days post-MI, myeloid cell depletion attenuated vascular accumulation of Nox2+CD45+ cells, endothelial dysfunction, oxidative stress, and vascular expression of adhesion molecules and angiotensin II receptor type 1 (AT1R). Pharmacological blockade of this receptor for 4 weeks did not significantly alter cardiac function, but mimicked the effects of myeloid cell depletion: telmisartan (20 mg/kg/day, fed to C57BL/6J mice) diminished bone marrow myelopoesis and myeloid reactive oxygen species production, attenuated endothelial leucocyte rolling and vascular accumulation of CD11b+Ly6G-Ly6Chigh monocytes and macrophages, resulting in improved vascular function with less abundance of Nox2+CD45+ cells. Endothelial dysfunction in HF ensuing MI is mediated by inflammatory Nox2+ myeloid cells infiltrating the vessel wall that can be targeted by AT1R blockade.
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)
Sigma-Aldrich
Histopaque®-1119, sterile-filtered, density: 1.119