MilliporeSigma
  • Voltage-gated proton channels from fungi highlight role of peripheral regions in channel activation.
Voltage-gated proton channels from fungi highlight role of peripheral regions in channel activation.
Communications biology (2021-02-28)
Chang Zhao, Francesco Tombola
ABSTRACT
Here, we report the identification and characterization of the first proton channels from fungi. The fungal proteins are related to animal voltage-gated Hv channels and are conserved in both higher and lower fungi. Channels from Basidiomycota and Ascomycota appear to be evolutionally and functionally distinct. Representatives from the two phyla share several features with their animal counterparts, including structural organization and strong proton selectivity, but they differ from each other and from animal Hvs in terms of voltage range of activation, pharmacology, and pH sensitivity. The activation gate of Hv channels is believed to be contained within the transmembrane core of the protein and little is known about contributions of peripheral regions to the activation mechanism. Using a chimeragenesis approach, we find that intra- and extracellular peripheral regions are main determinants of the voltage range of activation in fungal channels, highlighting the role of these overlooked components in channel gating.
Sigma-Aldrich
2-Guanidinobenzimidazole, 95%