MilliporeSigma
  • Ceramide kinase mediates intrinsic resistance and inferior response to chemotherapy in triple-negative breast cancer by upregulating Ras/ERK and PI3K/Akt pathways.
Ceramide kinase mediates intrinsic resistance and inferior response to chemotherapy in triple-negative breast cancer by upregulating Ras/ERK and PI3K/Akt pathways.
Cancer cell international (2021-01-13)
Shan Zhu, Yulin Xu, Lijun Wang, Shichong Liao, Yuan Wang, Manman Shi, Yi Tu, Yurong Zhou, Wen Wei
ABSTRACT
Clinical management of triple-negative breast cancer (TNBC) patients remain challenging because of the development of chemo-resistance. Identification of biomarkers for risk stratification of chemo-resistance and therapeutic decision-making to overcome such resistance is thus necessary. Retrospective analysis was performed to identify potential stratification biomarkers. The levels of ceramide kinase (CERK) was determined in breast cancer patients. The roles of CERK and its downstream signaling pathways were analysed using cellular and biochemical assays. CERK upregulation was identified as a biomarker for chemotherapeutic response in TNBC. A > 2-fold change in CERK (from tumor)/CERK (from normal counterpart) was significantly associated with chemo-resistance (OR = 2.66, 95% CI 1.18-7.34), P = 0.04. CERK overexpression was sufficient to promote TNBC growth and migration, and confer chemo-resistance in TNBC cell lines, although this resistance could be overcome via CERK inhibition. Mechanistic studies suggest that CERK mediates intrinsic resistance and inferior response to chemotherapy in TNBC by regulating multiple oncogenic pathways such as Ras/ERK, PI3K/Akt/mTOR, and RhoA. Our work provides an explanation for the heterogeneity of chemo-response across TNBC patients and demonstrates that CERK inhibition offers a therapeutic strategy to overcome treatment resistance.
Sigma-Aldrich
MISSION® esiRNA, targeting human CERK
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder