MilliporeSigma
  • SRPS associated protein WDR60 regulates the multipolar-to-bipolar transition of migrating neurons during cortical development.
SRPS associated protein WDR60 regulates the multipolar-to-bipolar transition of migrating neurons during cortical development.
Cell death & disease (2021-01-14)
Cui Li, Yu Zheng, Yufang Zheng, Zhiheng Xu
ABSTRACT
Mutations of WD40 repeat domain 60 (WDR60) have been identified in short-rib polydactyly syndromes (SRPS I-V), a group of lethal congenital disorders characterized by short ribs, polydactyly, and a range of extraskeletal phenotypes. However, the underlying mechanism is still unclear. Here, we report that WDR60 is essential for embryonic development and plays a critical role in the multipolar-bipolar transition and migration of newborn neurons during brain development. Mechanically, we found that WDR60 was located at the microtubule-organizing center to control microtubule organization and possibly, the trafficking of cellular components. Importantly, the migration defect caused by Wdr60 knockdown could be rescued by the stable form of α-Tubulin, α-TubulinK40Q (an acetylation-mimicking mutant). These findings identified a non-cilia function of WDR60 and provided insight into its biological function, as well as the pathogenesis of WDR60 deficiency associated with SRPS.
Sigma-Aldrich
Anti-WDR60 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1