MilliporeSigma
  • A novel potent inhibitor of 2-amino-1-methyl-6-phenylimidazo[4,5-b] pyridine (PhIP) formation from Chinese chive: Identification, inhibitory effect and action mechanism.
A novel potent inhibitor of 2-amino-1-methyl-6-phenylimidazo[4,5-b] pyridine (PhIP) formation from Chinese chive: Identification, inhibitory effect and action mechanism.
Food chemistry (2020-12-11)
Qi Wang, Weiwei Cheng, Yifeng Zhang, Qingzheng Kang, Vemana Gowd, Yuanyuan Ren, Feng Chen, Ka-Wing Cheng
ABSTRACT
Differential solvent extraction and phytochemical profiling of Chinse chive were employed to identify its principal PhIP-formation inhibitory constituents. Six compounds (mangiferin, isorhamnetin, luteolin, rosmarinic acid, 6-methylcoumarin, and cyanidin-3-glucoside) were further analyzed in a PhIP-producing chemical model to identify the dominant inhibitor. Its inhibitory mechanism was investigated by assessing the contribution of antioxidation and scavenging of key PhIP precursor/intermediate. No significant correlation was observed between PhIP inhibition rates and antioxidant activities. Further evaluation of the novel potent inhibitor mangiferin revealed a highly significant correlation between its dose-dependent inhibition of PhIP formation and phenylacetaldehyde scavenging. Finally, the proposed mechanism was corroborated through organic synthesis and structural elucidation of the mangiferin-phenylacetaldehyde adduct. This study has identified a potent novel inhibitor of the most abundant HA in heat-processed food and characterized its action mechanism. These findings may provide insight for future studies on mitigation of dietary exposure to toxic Maillard products by polyphenolic phytochemicals.
Sigma-Aldrich
Butyl methyl ether, 99%
Sigma-Aldrich
(±)-6-Hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid, 97%