MilliporeSigma
  • Polymer-Nematic Liquid Crystal Interface: On the Role of the Liquid Crystalline Molecular Structure and the Phase Sequence in Photoalignment.
Polymer-Nematic Liquid Crystal Interface: On the Role of the Liquid Crystalline Molecular Structure and the Phase Sequence in Photoalignment.
Polymers (2021-01-13)
Ameer R K Nassrah, István Jánossy, Viktor Kenderesi, Tibor Tóth-Katona
ABSTRACT
We provide experimental evidence for the influence of the molecular structure of the nematic liquid crystal (NLC) on the photoalignment process in three dimensions at the interface with a polymer layer. In particular, the experimental findings are explained through the presence (or absence) of the π-π aromatic interactions between the NLC and the polymer. The influence of the nematic-to-smectic A phase transition on the photocontrol is also addressed. Furthermore, we demonstrate that the photo-induced reorientation scenarios can be eventually connected to conformational changes in the photosensitive polymer.
Sigma-Aldrich
1-(trans-4-Hexylcyclohexyl)-4-isothiocyanatobenzene, liquid crystal (nematic), 99%